Remdesivir
Bottle, 50 mg
Catalog # APOSBISN0317-50MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Remdesivir is an nucleotide-analog prodrug with broad spectrum antiviral activity against RNA viruses. Remdesivir has demonstrated in vitro and in vivo activity in animal models against the viral pathogens MERS and SARS
Synonyms: 2-Ethylbutyl ((S)-(((2R,3S,4R,5R)-5-(4-aminopyrrolo[2,1-f][1,2,4]triazin-7-yl)-5-cyano-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(phenoxy)phosphoryl)-L-alaninate
- CAS Number:1809249-37-3
- Formula:C₂₇H₃₅N₆O₈P.CH₃SO₃H
- MDL Number:MFCD31657351
- Molecular Weight:698.682 g/mol
- Storage Temperature:Køleskab
Specifications
Specifications
- Pakning:Bottle
- Pakn:50 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



