PP242 free base ≥99%
Bottle, 5 mg
APOSBIFK0004-5MG
: 2-(4-Amino-1-isopropyl-1H-pyrazolo[3,4-d]pyrimidin-3-yl)-1H-indol-5-ol, Torkinib, 2-[4-Amino-1-(1-methylethyl)-1H-pyrazolo[3,4-d]pyrimidin-3-yl]-1H-indol-5-ol
- :1092351-67-1
- :C₁₆H₁₆N₆O
- :MFCD12196869
- :308.3 g/mol
- :146…156 °C
- :Fryser
- Pakning:Bottle
- Pakn:5 mg