Celite® 545, GPR RECTAPUR®
HDPE bottle, 250 g
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
Diatomaceous silica for filtration.
Celite® is used as a filtering aid or as adsorbent in column chromatography.
Synonymer: DE, Diatomaceous earth, flux-calcined
- CAS nummer:68855-54-9
- Formel:SiO₂
- MDL nummer:MFCD00132803
- Molecular Weight:60.08 g/mol
Specifikationer
Specifikationstestresultater
- Identification:Passes test
- pH (25°C; 10%):≥ 8,5
- Loss on ignition (600 °C):≤ 0.6%
- Particle size (< 0.1 mm):≥ 60%
Specifikationer
- Pakning:HDPE bottle
- Pakn:250 g



