Orders LinkContactUsLinkComponent
Lids for Clifton Baths
Lids for Clifton Baths
Clear plastic lift-off gabled lid designed to allow full use of the whole width of a Clifton water bath with the lowest height of the gable being 45 mm above the bath. This means test tube racks or flasks can be placed across the whole width of the bath without fouling the lid, maximising the bath working area. Insulated handle. Lid fits into the tank ensuring all condensation runs back into the bath and not down the side onto the bench top., NE2-10D, NE1-8, NE2D-4/8, NE2-8D, NE5-10D, NE2D-8/22 and NE1D-8
Katalog # NICKGL1-8AF
Leverandør:   NICKEL ELECTRO
Lids for Clifton Baths
Katalog # NICKGL1-8AF
Leverandør:  NICKEL ELECTRO
Lids for Clifton Baths
Lids for Clifton Baths
Clear plastic lift-off gabled lid designed to allow full use of the whole width of a Clifton water bath with the lowest height of the gable being 45 mm above the bath. This means test tube racks or flasks can be placed across the whole width of the bath without fouling the lid, maximising the bath working area. Insulated handle. Lid fits into the tank ensuring all condensation runs back into the bath and not down the side onto the bench top., NE2-10D, NE1-8, NE2D-4/8, NE2-8D, NE5-10D, NE2D-8/22 and NE1D-8
Price
1.380,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Clear plastic lift-off gabled lid designed to allow full use of the whole width of a Clifton water bath with the lowest height of the gable being 45 mm above the bath. This means test tube racks or flasks can be placed across the whole width of the bath without fouling the lid, maximising the bath working area. Insulated handle. Lid fits into the tank ensuring all condensation runs back into the bath and not down the side onto the bench top.
  • Seq:
    13
  • Til:
    NE2-10D, NE1-8, NE2D-4/8, NE2-8D, NE5-10D, NE2D-8/22 and NE1D-8