Accessories for Ebro Data Loggers
EBI 12-T222 Temperature logger, 1-channel, radial, 40 mm rigid metal probe
Catalog # EBRO1340-6610
Supplier: EBRO
Jump to:
Product Details & Documents
Documents
Data Loggers - On The Safe Side with Validated Processes
Application Notes
EBI 300/310/330 USB Data Loggers for Cold Chain Monitering.pdf Pasteurization Control Testing and Storage Life of Food.pdf USB Dataloggers for Cold Chain Monitoring.pdf Validation and Process Monitoring EBI11-12.pdf Validation and Process Monitoring Food.pdf
Additional Product Details
More
Manual Data Logger EBI 12 EX Series Manual Interface.pdf Manual USB Dataloggers for Cold Chain Monitoring.pdf
Manual
Professional Dataloggers
Product Literature
Specifications
- Beskrivelse:EBI 12-T222 Temperature logger, 1-channel, radial, 40 mm rigid metal probe
- Seq:34