Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant PYM (from E. coli)

Human Recombinant PYM (from E. coli)

Katalog # ABCAAB98083-50
Leverandør:   Abcam
Human Recombinant PYM (from E. coli)
Katalog # ABCAAB98083-50
Leverandør:  Abcam
CAS nummer:  
Human Recombinant PYM (from E. coli)
Human Recombinant PYM (from E. coli)
CAS nummer:  
Price
4.080,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant Human PYM protein is a Human Full Length protein, in the 1 to 204 aa range, expressed in Escherichia coli, with ≥85% purity and suitable for SDS-PAGE.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Protein/peptide name:
    PYM
  • Protein synonyms:
    PYM|WIBG|PYM1|Partner of Y14 and mago|PYM homolog 1 exon junction complex-associated factor|Protein wibg homolog
  • Biologisk aktivitet:
    Standard
  • Koncentration:
    0,25 mg/ml
  • Dyrefri:
    N
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB98083-50
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 8, Constituents: 20% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,00174% PMSF
  • Renhed:
    ≥85%
  • Tested applications:
    SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at −20 °C

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.