Human Recombinant PKM2 (from E. coli)
Product Details & Documents
Recombinant human PKM2 protein (Active) is a Human Full Length protein, in the 1 to 531 aa range, expressed in Escherichia coli, with >95% purity, <1 EU/µg endotoxin level and suitable for SDS-PAGE, WB, FuncS.
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifikationer
Specifikationer
- Konjugation:Unconjugated
- Form:Liquid
- Protein/peptide name:PKM2
- Protein synonyms:OIP3|PK2|PK3|PKM|Pyruvate kinase PKM|Cytosolic thyroid hormone-binding protein|Opa-interacting protein 3|Pyruvate kinase 2/3|Pyruvate kinase muscle isozyme|Threonine-protein kinase PKM2|Thyroid hormone-binding protein 1|Tumor M2-PK|Tyrosine-protein kinase PKM2|p58|CTHBP|OIP-3|THBP1
- Biologisk aktivitet:Bioactive
- Koncentration:1 mg/ml
- Dyrefri:N
- Shipping temperature:20
- Cat. No.:ABCAAB89364-500
- Protein/peptide type:Recombinant
- Formulering:pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,316% Tris HCl
- Endotoksinindhold:<1 EU/µg
- Renhed:>95%
- Tested applications:FuncS, SDS-PAGE, WB
- Kilde:E. coli
- Species:Human
- Opbevaring betingelser:Store at −20 °C



