Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant NSE (from E. coli)

Human Recombinant NSE (from E. coli)

Katalog # ABCAAB78797-500
Marketsource
Leverandør:   Abcam
Human Recombinant NSE (from E. coli)
Katalog # ABCAAB78797-500
Leverandør:  Abcam
Marketsource
CAS nummer:  
Human Recombinant NSE (from E. coli)
Human Recombinant NSE (from E. coli)
CAS nummer:  
Price
12.540,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant Human NSE protein is a Human Full Length protein, expressed in Escherichia coli, with >95% purity, <1 EU/µg endotoxin level and suitable for SDS-PAGE, WB.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Protein/peptide name:
    NSE
  • Protein synonyms:
    Gamma-enolase|2-phospho-D-glycerate hydro-lyase|Enolase 2|Neural enolase|Neuron-specific enolase|ENO2
  • Biologisk aktivitet:
    Standard
  • Koncentration:
    1 mg/ml
  • Dyrefri:
    N
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB78797-500
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 7,5, Constituents: 0,75% Potassium chloride, 0,242% Tris, 0,06% Magnesium sulphate
  • Endotoksinindhold:
    <1 EU/µg
  • Renhed:
    >95%
  • Tested applications:
    SDS-PAGE, WB
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at –20 °C

Specifikationer