Human Granzyme B ELISPOT Set
Human Granzyme B ELISPOT set
Katalog # ABCAAB46581-10X96
Leverandør: Abcam
Gå til:
Product Details & Documents
Human Granzyme B ELISPOT Set is a ELISPOT Set for the sensitive detection of Human Granzyme B production in a single cell suspension.
Specifikationer
Specifikationer
- Beskrivelse:Human Granzyme B ELISPOT set
- Cat. No.:ABCAAB46581-10X96
- Form:Uncoated
- Species reactivity:Human
- Processing time:Multiple steps
- Prøvetype:Suspension cells
- Opbevaringstemperatur:+4 °C
- Regulatory status:RUO
- Antigen name:Granzyme B
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



