Orders LinkContactUsLinkComponent
Multiple affinity removal starter reagent kits
Multiple affinity removal starter reagent kits
Multiple affinity removal reagent kit, -, 1 kt
Katalog # AGLS5185-5986
Leverandør:   AGILENT
Multiple affinity removal starter reagent kits
Katalog # AGLS5185-5986
Leverandør:  AGILENT
CAS nummer:  
Multiple affinity removal starter reagent kits
Multiple affinity removal starter reagent kits
Multiple affinity removal reagent kit, -, 1 kt
CAS nummer:  
Price
5.895,00 kr
Log ind for tilgængelighed
1 * 1 kt
Product Details & Documents
Starter reagent kit for multiple affinity removal LC columns.
Leveringsinformation: Includes: 2 bottles buffer A (1 L), 1 bottle buffer B (1 L), 2 packs of 25 spin filters, 1 pack of 25 spin concentrator.
Specifikationer
Specifikationer
  • Pakn:
    1 kt
  • Beskrivelse:
    Multiple affinity removal reagent kit
  • Details:
    -