Polypropylene Muff Covers with Elastic
Polypropylene muff cover, 9" (229 mm), fits 4 - 5,5" (102 - 140 mm) Ø, hvid
Katalog # 129-3072
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Gå til:
Product Details & Documents
Keeps cleaned non sterile production hoses protected from external sources of contamination.
- Manufactured and packed in a controlled cleanroom environment
- Reduces labour costs
- Tear and puncture resistant
- Conveniently packaged in 100 per sealed poly bag
These polypropylene muffs are a great choice in non sterile pharmaceutical manufacturing operations. Can be used as a clean equipment cover to protect process hoses.
Poly bagged in packs of 100 to help ensure cleanliness up until point of use.
Leveringsinformation: Certificates of Conformance sent with each shipment.
Specifikationer
- Farve:hvid
- Beskrivelse:Polypropylene muff cover, 9" (229 mm), fits 4 - 5,5" (102 - 140 mm) Ø
- Seq:6