Polypropylene Muff Covers with Elastic
Polypropylene muff cover, 5" (127 mm), fits 1,75 - 2,5" (44,5 - 63,5 mm) Ø, hvid
Katalog # 129-3068
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Gå til:
Product Details & Documents
Keeps cleaned non sterile production hoses protected from external sources of contamination.
- Manufactured and packed in a controlled cleanroom environment
- Reduces labour costs
- Tear and puncture resistant
- Conveniently packaged in 100 per sealed poly bag
These polypropylene muffs are a great choice in non sterile pharmaceutical manufacturing operations. Can be used as a clean equipment cover to protect process hoses.
Poly bagged in packs of 100 to help ensure cleanliness up until point of use.
Leveringsinformation: Certificates of Conformance sent with each shipment.
Specifikationer
- Farve:hvid
- Beskrivelse:Polypropylene muff cover, 5" (127 mm), fits 1,75 - 2,5" (44,5 - 63,5 mm) Ø
- Seq:2