b.safe Funnels 250
Funnel, G2"+Trisure2"
Katalog # 590-0404
Leverandør: Bohlender
Gå til:
Product Details & Documents
Funnel made of PE for liquid waste disposal. The funnel can be safely mounted on the waste container with its threaded connection that is safely fixed with the filling neck.
- Hinged lid: With grip for easy opening and closing
- Sieve: Big grip for safe grapping and removal of the sieve
- Plug-in connection for filling tube: An additional filling tube can be plugged-in the funnel neck
A stainless steel sieve on the outlet restrains magnetic stirrer bars and other solid materials. It has a grip for easy removal for cleaning. The hinged lid reliably withholds vapours from the container.
Documents
Manual b.safe Funnels
Manuel
Specifikationer
- Gevind:G2"+Trisure2"
- Beskrivelse:Funnel
- Seq:02
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Sieve for funnel 180