Grounding cables, b.safe
Grounding cable with 2× ring cable lug M6, 1,5 m
Katalog # 590-0343
Leverandør: Bohlender
Gå til:
Product Details & Documents
For earth connection of vessels and components made of static dissipative materials.
- Choose the suitable connection to earth that we offer in our range of grounding cables with different connectors
- Customised versions can be offered on request
- Suitable grounding accessories for b.safe products
The scope of delivery of the ring cable lug includes screw, nut and washer.
Specifikationer
- Beskrivelse:Grounding cable with 2× ring cable lug M6
- Længde:1,5 m
- Seq:02