Orders LinkContactUsLinkComponent
Capillary cutter, b.safe
Capillary cutter, b.safe
Capillary cutter with guide bores for tubing up to Ext.Ø 8 mm
Catalog # 590-0334
Supplier:   Bohlender
Capillary cutter, b.safe
Catalog # 590-0334
Supplier:  Bohlender
Capillary cutter, b.safe
Capillary cutter, b.safe
Capillary cutter with guide bores for tubing up to Ext.Ø 8 mm
Price
656,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Ideal for cutting capillaries and elastic tubes without reinforcement.
  • Capillary cutter for up to tubing Ext.Ø 8 mm with guide bores
  • High qualitiy capillary cutter
  • Replacement blades available
The guide bores help to get a clean cut of the capillaries.
Specifications
  • Beskrivelse:
    Capillary cutter with guide bores for tubing up to Ext.Ø 8 mm
  • Seq:
    01