Spouts, b.safe
Spout tube flexible, S60/61, PE
590-0324
Screw cap for canister thread, freely turnable spout tube, rigid or flexible design with integrated safety aeration for constant flow.
- Spout tube is freely turnable
- Integrated safety aeration
- Prevents loss of fluids
Quick refilling of liquids without dripping directly from canister.
- Gevind:S60/61
- Beskrivelse:Spout tube flexible
- Materiale:PE
- Seq:03