Orders LinkContactUsLinkComponent
Laboratory bottles, b.safe
Laboratory bottles, b.safe
1000 ml, GLS 80
Catalog # 590-0286
Supplier:   Bohlender
Laboratory bottles, b.safe
Catalog # 590-0286
Supplier:  Bohlender
Laboratory bottles, b.safe
Laboratory bottles, b.safe
1000 ml, GLS 80
Price
390,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Laboratory flask made of Borosilicate glass including screw cap and spout made of PP.
  • High-quality borosilicate glass
  • Including screw cap
  • Including spout made of PP
b.safe offers also suitable containers like flasks and canisters in addition to the wide range of caps for the extraction of solvents and waste caps or funnels for the collection of liquid waste.
Unless otherwise indicated, all b.safe canisters are supplied with UN-approval and can be used for the transport of hazardous goods.
Packaging: Packing unit: 1 piece.
Specifications
  • Gevind:
    GLS 80
  • Kapacitet:
    1000 ml
  • Seq:
    02