Accessories for REVOLVE
Stage insert
Catalog # ECHLHLDR0007
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:Stage insert
- Seq:09
- Kontrastteknik:Lysfelt og fasekontrast
- Cat. No.:ECHLHLDR0007
- Output:USB
- Kamera:80 MP colour
- Elektriske specifikationer:168 - 240 VA (50/60 Hz)
- Display:12,9” iPad Pro® Retina® display, 2732×2048 (264 pixels/inch)
- Sensor:CMOS