Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Phthalsyre 98%

Phthalsyre 98%

Bottle, 100 g
Katalog # APOSOR923932-100G
Leverandør:   Apollo Scientific
Phthalsyre 98%
Katalog # APOSOR923932-100G
Leverandør:  Apollo Scientific
CAS nummer:  88-99-3
Phthalsyre 98%

Bottle, 100 g

CAS nummer:  88-99-3
Price
165,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  Pathalic acid
  • CAS nummer:88-99-3
  • Formel:C₈H₆O₄
  • MDL nummer:MFCD00002467
  • Molecular Weight:166.13 g/mol
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    100 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.