Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
4-Ethylpyrocatechol 97%

4-Ethylpyrocatechol 97%

Bottle, 5 g
Katalog # APOSOR925237-5G
Leverandør:   Apollo Scientific
4-Ethylpyrocatechol 97%
Katalog # APOSOR925237-5G
Leverandør:  Apollo Scientific
CAS nummer:  1124-39-6
4-Ethylpyrocatechol 97%

Bottle, 5 g

CAS nummer:  1124-39-6
Price
889,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  4-Ethylbenzene-1,2-diol, 4-Ethylcatechol
  • CAS nummer:1124-39-6
  • Formel:C₈H₁₀O₂
  • MDL nummer:MFCD00015847
  • Molecular Weight:138.17 g/mol
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    5 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.