Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
3,5-Diethyl-4-hydroxybenzaldehyde 95%

3,5-Diethyl-4-hydroxybenzaldehyde 95%

Bottle, 5 g
Katalog # APOSOR919394-5G
Leverandør:   Apollo Scientific
3,5-Diethyl-4-hydroxybenzaldehyde 95%
Katalog # APOSOR919394-5G
Leverandør:  Apollo Scientific
CAS nummer:  69574-07-8
3,5-Diethyl-4-hydroxybenzaldehyde 95%

Bottle, 5 g

CAS nummer:  69574-07-8
Price
7.270,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
  • CAS nummer:69574-07-8
  • Formel:C₁₁H₁₄O₂
  • MDL nummer:MFCD13152308
  • Molecular Weight:178.228 g/mol
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    5 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.