Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
3-Hydroxybenzonitril

3-Hydroxybenzonitril

Bottle, 1 g
Catalog # APOSOR6704-1G
Irritant
Supplier:   Apollo Scientific
3-Hydroxybenzonitril
Catalog # APOSOR6704-1G
Supplier:  Apollo Scientific
CAS Number:  873-62-1
3-Hydroxybenzonitril

Bottle, 1 g

CAS Number:  873-62-1
Price
165,00 kr
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  m-Hydroxybenzonitrile, m-Cyanophenol, 3-Cyanophenol
  • CAS Number:873-62-1
  • Formula:C₇H₅NO
  • MDL Number:MFCD00002252
  • Molecular Weight:119.12 g/mol
  • Melting Pt:78…81 °C
  • Storage Temperature:Stue temperatur
  • EINECS:212-845-4
Specifications

Specifications

  • Pakning:
    Bottle
  • Pakn:
    1 g

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.