Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
4-Ethylanisol 98%

4-Ethylanisol 98%

Bottle, 25 g
Katalog # APOSOR916875-25G
Leverandør:   Apollo Scientific
4-Ethylanisol 98%
Katalog # APOSOR916875-25G
Leverandør:  Apollo Scientific
CAS nummer:  1515-95-3
4-Ethylanisol 98%

Bottle, 25 g

CAS nummer:  1515-95-3
Price
231,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  1-Ethyl-4-methoxybenzene
  • CAS nummer:1515-95-3
  • Formel:C₉H₁₂O
  • MDL nummer:MFCD00015176
  • Molecular Weight:136.19 g/mol
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    25 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.