Orders LinkContactUsLinkComponent
Poly(vinylidene fluoride)
Poly(vinylidene fluoride)
Bottle, 250 g
Catalog # APOSPC6267-250G
Irritant
Supplier:   Apollo Scientific
Poly(vinylidene fluoride)
Catalog # APOSPC6267-250G
Supplier:  Apollo Scientific
CAS Number:  24937-79-9
Poly(vinylidene fluoride)
Bottle, 250 g
CAS Number:  24937-79-9
Price
933,00 kr
Log in for availability
1 * 250 g
Product Details & Documents
  • CAS Number:24937-79-9
  • Formula:(-CH₂CF₂-)n
  • MDL Number:MFCD00084470
  • Melting Pt:155…165 °C
  • Density:1,74 g/cm³ (25 °C)
  • Storage Temperature:Stue temperatur
Specifications
Specifications
  • Pakning:
    Bottle
  • Pakn:
    250 g
Specifications