Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

3-Hydroxy-5-methylbenzonitrile 98%

Bottle, 250 mg
Catalog # APOSOR928299-250MG
Supplier:   Apollo Scientific
3-Hydroxy-5-methylbenzonitrile 98%
Catalog # APOSOR928299-250MG
Supplier:  Apollo Scientific
CAS Number:  95658-81-4
3-Hydroxy-5-methylbenzonitrile 98%

Bottle, 250 mg

CAS Number:  95658-81-4
Price
736,00 kr
Log in for availability
1 * 250 mg
Product Details & Documents
  • CAS Number:95658-81-4
  • Formula:C8H7NO
  • MDL Number:MFCD16999137
  • Molecular Weight:133.1500 g/mol
  • Storage Temperature:Stue temperatur
Specifications

Specifications

  • Pakning:
    Bottle
  • Pakn:
    250 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.