Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Thiomorpholin 98%

Bottle, 5 g
Catalog # APOSOR6523-5G
Corrosive
Supplier:   Apollo Scientific
Thiomorpholin 98%
Catalog # APOSOR6523-5G
Supplier:  Apollo Scientific
CAS Number:  123-90-0
Thiomorpholin 98%

Bottle, 5 g

CAS Number:  123-90-0
Price
165,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:123-90-0
  • Formula:C₄H₉NS
  • MDL Number:MFCD00005974
  • Molecular Weight:103.19 g/mol
  • Boiling Pt:169…177 °C
  • Density:1,026 g/cm³ (25 °C)
  • Flash Pt:67 °C
  • Storage Temperature:Køleskab
  • EINECS:204-660-2
  • UN:3267
  • ADR:8,III
Specifications

Specifications

  • Pakning:
    Bottle
  • Pakn:
    5 g

Specifications