Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

2,5-Dimethoxypyridine 95%

Bottle, 5 g
Catalog # APOSOR471688-5G
Supplier:   Apollo Scientific
2,5-Dimethoxypyridine 95%
Catalog # APOSOR471688-5G
Supplier:  Apollo Scientific
CAS Number:  867267-24-1
2,5-Dimethoxypyridine 95%

Bottle, 5 g

CAS Number:  867267-24-1
Price
286,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:867267-24-1
  • Formula:C₇H₉NO₂
  • MDL Number:MFCD12546620
  • Molecular Weight:139.152 g/mol
  • Storage Temperature:Stue temperatur
Specifications

Specifications

  • Pakning:
    Bottle
  • Pakn:
    5 g

Specifications