Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

2,2'-Difluorbiphenyl

Bottle, 5 g
Katalog # APOSPC2748-5G
Irriterende
Leverandør:   Apollo Scientific
2,2'-Difluorbiphenyl
Katalog # APOSPC2748-5G
Leverandør:  Apollo Scientific
CAS nummer:  388-82-9
2,2'-Difluorbiphenyl

Bottle, 5 g

CAS nummer:  388-82-9
Price
2.020,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
  • CAS nummer:388-82-9
  • Formel:C₁₂H₈F₂
  • Molecular Weight:190.19 g/mol
  • Smeltende Pt:117…121 °C
  • ADR:9,II
  •  :Density: 1.393 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • FN:3152
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    5 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.