(3-Fluorophenoxy)acetic acid
Bottle, 5 g
Katalog # APOSPC200534-5G
Leverandør: Apollo Scientific
(3-Fluorophenoxy)acetic acid
Katalog # APOSPC200534-5G
Leverandør: Apollo Scientific
CAS nummer: 404-98-8
Gå til:
Product Details & Documents
Synonymer: 2-(3-Fluorophenoxy)acetic acid
- CAS nummer:404-98-8
- Formel:C₈H₇FO₃
- MDL nummer:MFCD02295721
- Molecular Weight:170.14 g/mol
- Opbevaringstemperatur:Stue temperatur
Specifikationer
Specifikationer
- Pakning:Bottle
- Pakn:5 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



