Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Biohazard Waste Containers

Biohazard Waste Containers

rød, 34 L, 354×464 mm
Katalog # JUSU05930RD
Leverandør:   JUSTRITE MANUFACTURING
Biohazard Waste Containers
Katalog # JUSU05930RD
Leverandør:  JUSTRITE MANUFACTURING
Biohazard Waste Containers
Biohazard Waste Containers

rød, 34 L, 354×464 mm

Price
1.360,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

These leakproof sturdy steel containers with foot-operated and self-closing lid may be used to store contaminated laundry as well as other regulated waste (not for sharps). They have a biohazard bag which is held securely around the circumference of the can top.

 : Comply with US OSHA Standard 29 CFR Part 1910.1030 for segregation of infectious waste. FM and TUV approved.

Specifikationer
  • Diameter×H:
    354×464 mm
  • Kapacitet:
    34 L
  • Farve:
    rød
  • Seq:
    03
Accessories

Produktinformation

  • Katalog #
    Farve
    Kapacitet
    Tilgængelighed
    Price
  • Biohazard waste can liner bags
    Biohazard waste can liner bags
    JUSU05901
    rød
    56,8 L
    Log ind for tilgængelighed
    1 * 100 Items - 214,00 kr