0,00 kr
Ordreindtastning
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
Orders Link
ContactUsLinkComponent
Mikroskoper
Condensers for REVOLVE 3M microscope
Condensers for REVOLVE 3M microscope
ELWD universal condenser, WD 73 mm, NA 0,30 aperture
630-3323
:
DISCOVER ECHO
Condensers for REVOLVE 3M microscope
630-3323
:
DISCOVER ECHO
Condensers for REVOLVE 3M microscope
ELWD universal condenser, WD 73 mm, NA 0,30 aperture
11.980,00 kr
1 * 1 Items
-
+
ECHO Meet Revolve
Beskrivelse:
ELWD universal condenser, WD 73 mm, NA 0,30 aperture
Seq:
01
Onsite install and training
#: 630-3343
: DISCOVER ECHO