Objectives for REVOLVE 3M microscope
Plan achromat (N-plan) oil phase Ph3; 1,25 NA, 100×, 0,15 mm
Katalog # ECHLJMAG1009
Leverandør: DISCOVER ECHO
Gå til:
Product Details & Documents
Documents
ECHO Meet Revolve
Produktlitteratur
Specifikationer
- Forstørrelse:100×
- Beskrivelse:Plan achromat (N-plan) oil phase Ph3; 1,25 NA
- Seq:28
- Arbejdsafstand:0,15 mm
- Cat. No.:ECHLJMAG1009