Anisol ≥99%, GPR RECTAPUR®
Plastic jerrican, 25 L
: Methyl phenyl ether, Methoxybenzene
- :100-66-3
- :C₇H₈O
- :MFCD00008354
- :108.14 g/mol
- :52 °C
- :Stue temperatur
- :2222
- :3,III
- Assay :≥ 99 %
- Appearance :Clear colourless liquid
- Odour:Characteristic
- Density (20/4) :0,992 - 0,994
- Pakning:Plastic jerrican
- Pakn:25 L



