Anisol ≥99%, GPR RECTAPUR®
Glass bottle, 2,5 L
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
Synonymer: Methyl phenyl ether, Methoxybenzene
- CAS nummer:100-66-3
- Formel:C₇H₈O
- MDL nummer:MFCD00008354
- Molecular Weight:108.14 g/mol
- Flash Pt:52 °C
- Opbevaringstemperatur:Stue temperatur
- FN:2222
- ADR:3,III
Specifikationer
Specifikationstestresultater
- Assay :≥ 99 %
- Appearance :Clear colourless liquid
- Odour:Characteristic
- Density (20/4) :0,992 - 0,994
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L



