Anisol ≥99%, GPR RECTAPUR®
Glass bottle, 1 L
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
Synonymer: Methyl phenyl ether, Methoxybenzene
- CAS nummer:100-66-3
- Formel:C₇H₈O
- MDL nummer:MFCD00008354
- Molecular Weight:108.14 g/mol
- Flash Pt:52 °C
- Opbevaringstemperatur:Stue temperatur
- FN:2222
- ADR:3,III
Documents
Certifikater
Søg efter partinummerP20_VWRC87106.290_SPEC_EN.pdf spec_VWRC87106.290 spec_VWRC87106.320
Produktspecifikation
Specifikationer
Specifikationstestresultater
- Assay :≥ 99 %
- Appearance :Clear colourless liquid
- Odour:Characteristic
- Density (20/4) :0,992 - 0,994
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



