Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Glass beads

Glass beads

Glass beads in glass bottle, 5,0 mm
Katalog # ACRO465951000
Leverandør:   Thermo Fisher Scientific
Glass beads
Katalog # ACRO465951000
Glass beads

Glass beads in glass bottle, 5,0 mm

Price
192,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

MDL No.: MFCD00131156 Appearance: Glass beads Storage Temperature: Ambient

Documents

Specifikationer
  • Ø:
    5,0 mm
  • Beskrivelse:
    Glass beads in glass bottle
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.