Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Glass beads

Glass beads in glass bottle, 1,0 mm
Katalog # ACRO465941000
Leverandør:   Thermo Fisher Scientific
Glass beads
Katalog # ACRO465941000
Glass beads

Glass beads in glass bottle, 1,0 mm

Price
192,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

MDL No.: MFCD00131156 Appearance: Glass beads Storage Temperature: Ambient

Specifikationer
  • Ø:
    1,0 mm
  • Beskrivelse:
    Glass beads in glass bottle