Orders LinkContactUsLinkComponent
[EN]NEXION 5000 SETUP SOLUTION 1% HNO3, 1 * 1 Items
[EN]NEXION 5000 SETUP SOLUTION 1% HNO3, 1 * 1 Items
Katalog # PERCN8162301
Leverandør:   PerkinElmer
[EN]NEXION 5000 SETUP SOLUTION 1% HNO3, 1 * 1 Items
Katalog # PERCN8162301
Leverandør:  PerkinElmer
[EN]NEXION 5000 SETUP SOLUTION 1% HNO3, 1 * 1 Items
Price
1.700,00 kr