Orders LinkContactUsLinkComponent
CloneMatrix
CloneMatrix
CloneMatrix CHO (1×40 ml), 1 Bottle
Katalog # MLDVK8530
Leverandør:   Molecular Devices
CloneMatrix
Katalog # MLDVK8530
Leverandør:  Molecular Devices
CAS nummer:  
CloneMatrix
CloneMatrix CHO (1×40 ml), 1 Bottle
CAS nummer:  
Price
4.760,00 kr
Log ind for tilgængelighed
1 * 1 Fl
Product Details & Documents
CloneMatrix is a 2,5X concentrate that serves as a matrix to produce semi-solid media for hybridoma, myeloma, CHO, HEK, and mESC.
Specifikationer
Specifikationer
  • Pakn:
    1 Bottle
  • Beskrivelse:
    CloneMatrix CHO (1×40 ml)