Orders LinkContactUsLinkComponent
Beakers, with handle, PP
Beakers, with handle, PP
5000 ml, 257 mm, 203 mm, 100 ml
Katalog # BURK7204-5000
Leverandør:   BURKLE
Beakers, with handle, PP
Katalog # BURK7204-5000
Leverandør:  BURKLE
Beakers, with handle, PP
Beakers, with handle, PP
5000 ml, 257 mm, 203 mm, 100 ml
Price
195,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PP, highly transparent.
  • With embossed graduation printed in blue
  • Ergonomic open or close handle for secure grip
  • Functional spout to minimize spills
Ideal for industrial applications.
Specifikationer
  • Udv.Ø:
    203 mm
  • Inddelings-interval:
    100 ml
  • Kapacitet:
    5000 ml
  • Højde:
    257 mm
  • Seq:
    05