Orders LinkContactUsLinkComponent
Universal reference RNAs
Universal reference RNAs
Universal rat reference RNA, 2×200 µg, 2
Katalog # AGLS740200
Leverandør:   AGILENT
Universal reference RNAs
Katalog # AGLS740200
Leverandør:  AGILENT
CAS nummer:  
Universal reference RNAs
Universal rat reference RNA, 2×200 µg, 2
CAS nummer:  
Price
7.208,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents
High-quality total RNA for human, mouse, and rat microarray gene-expression profiling that acts as a consistent control for standard data set comparisons.
  • Comprised of 10 cell lines for human, 11 for mouse and 14 for rat giving broad gene coverage
  • UHRR selected as one of the two standards in the FDA led Microarray Quality Control (MAQC) project
The reference RNA is designed to be used as a reference for uarray gene-profiling experiments.
Delivery information: Includes RNase-free water, 1,5 ml.
Caution: For research use only. Not for use in diagnostic procedures.
Specifikationer
Specifikationer
  • Pakn:
    2
  • Beskrivelse:
    Universal rat reference RNA, 2×200 µg