Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Pfu DNA polymerase

Cloned Pfu DNA polymerase, Cloned Pfu DNA polymerase (2.5 U/µl), 10X cloned Pfu buffer, 500 U, 1 KIT
  AGLS600154
 :   AGILENT
Pfu DNA polymerase
  AGLS600154
 :  AGILENT
 :  
Pfu DNA polymerase

Cloned Pfu DNA polymerase, Cloned Pfu DNA polymerase (2.5 U/µl), 10X cloned Pfu buffer, 500 U, 1 KIT

 :  
 
5.525,00 kr
   
1 * 1 Each
 

High-quality polymerases for difficult to amplify DNA targets.

  • Proofreading DNA polymerase isolated from Pyrococcus furiosus
  • Ideal choice for many techniques requiring high-fidelity DNA synthesis by PCR
  • Blunt ends

Pfu DNA Polymerases includes Pfu DNA Polymerase, a proofreading DNA polymerase isolated from Pyrococcus furiosus.

The portfolio also features the recombinant Exo- Pfu DNA Polymerase (a genetically engineered mutant of the cloned Pfu DNA format) and the PfuTurbo DNA Polymerase with an optional alternative detergent.

 : For research use only. Not for use in diagnostic procedures.

 

 

  • Pakn:
    1 KIT
  • Beskrivelse:
    Cloned Pfu DNA polymerase
  • Størrelse:
    500 U
  • Details:
    Cloned Pfu DNA polymerase (2.5 U/µl), 10X cloned Pfu buffer