Pfu DNA polymerase
Pfu DNA polymerase, Native Pfu DNA polymerase (2.5 U/µl), 10X native Pfu buffer, 1000 U, 1
AGLS600140
: AGILENT
High-quality polymerases for difficult to amplify DNA targets.
- Proofreading DNA polymerase isolated from Pyrococcus furiosus
- Ideal choice for many techniques requiring high-fidelity DNA synthesis by PCR
- Blunt ends
Pfu DNA Polymerases includes Pfu DNA Polymerase, a proofreading DNA polymerase isolated from Pyrococcus furiosus.
The portfolio also features the recombinant Exo- Pfu DNA Polymerase (a genetically engineered mutant of the cloned Pfu DNA format) and the PfuTurbo DNA Polymerase with an optional alternative detergent.
: For research use only. Not for use in diagnostic procedures.
- Pakn:1
- Beskrivelse:Pfu DNA polymerase
- Størrelse:1000 U
- Details:Native Pfu DNA polymerase (2.5 U/µl), 10X native Pfu buffer



