Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Resin, StrataClean

Resin, StrataClean

StrataClean resin, 3 ml, 1
Katalog # AGLS400714
Leverandør:   AGILENT
Resin, StrataClean
Katalog # AGLS400714
Leverandør:  AGILENT
CAS nummer:  
Resin, StrataClean

StrataClean resin, 3 ml, 1

CAS nummer:  
Price
2.728,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

StrataClean resin eliminates the need to perform phenol:chloroform extractions, which are highly toxic and combustible.

  • Simple, fast, and easy to use protocol
  • Replaces toxic and tedious acetone precipitation, dialysis and ultrafiltration protocols to concentrate proteins

The resin is a non-toxic, non-combustible slurry of hydroxylated silica particles that is used to clean up DNA or concentrate dilute protein mixtures.

DNA extracted with StrataClean resin may be used directly for sequencing or can be precipitated and used for manipulations such as ligations and restriction digests.

Caution: For research use only. Not for use in diagnostic procedures.

Specifikationer

Specifikationer

  • Pakn:
    1
  • Beskrivelse:
    StrataClean resin, 3 ml
  • Cat. No.:
    AGLS400714
  • Prep size:
    Scalable
  • Refine target molecule:
    DNA preps|Total DNA|Amplified/modified DNA
  • Target molecule:
    DNA
  • Method / Format:
    Affinity resin
  • Sample type:
    DNA preps|PCR and other enzymatic reactions

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.