Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

CryoStor® CS10 Cryopreservation Medium

CryoStor® CS10, 160 ml (10 ml vials), Human, mouse, non human primate, other, rat; animal component-free, serum-free, 1 KIT
Katalog # STMC07952
Marketsource
Leverandør:   Stemcell Technologies
CryoStor® CS10 Cryopreservation Medium
Katalog # STMC07952
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
CryoStor® CS10 Cryopreservation Medium
CryoStor® CS10 Cryopreservation Medium

CryoStor® CS10, 160 ml (10 ml vials), Human, mouse, non human primate, other, rat; animal component-free, serum-free, 1 KIT

CAS nummer:  
Price
11.405,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Animal component-free, defined cryopreservation medium with 10% DMSO.

  • Ready-to-use
  • Serum-free and protein-free
  • Animal component-free
  • cGMP manufactured with USP grade/highest-quality components
  • FDA master file
  • Sterility, endotoxin, and cell-based quality control testing

Maximise post-thaw cell recovery and viability following cryopreservation at very low temperatures (–70 to –196 °C) with ready-to-use CryoStor® CS10 medium. Serum-free, animal component-free, and cGMP-manufactured, this defined medium provides a safe, protective environment that is recommended for the cryopreservation of a variety of sensitive cell and sample types, including myeloma cell lines, human pluripotent stem cells, blood-derived cells, and more. Available in a variety of convenient formats, CryoStor® CS10 is formulated with USP-grade components to minimise variability and contains 10% DMSO.

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    1 KIT
  • Beskrivelse:
    CryoStor® CS10, 160 ml (10 ml vials)
  • Udformning:
    Human, mouse, non human primate, other, rat; animal component-free, serum-free

Specifikationer