Orders LinkContactUsLinkComponent
Accessories for PLATEMASTER®
Accessories for PLATEMASTER®
384-well plate adapter, positioning by hand
Katalog # 631-1359
Leverandør:   GILSON
Accessories for PLATEMASTER®
Katalog # 631-1359
Leverandør:  GILSON
Accessories for PLATEMASTER®
Accessories for PLATEMASTER®
384-well plate adapter, positioning by hand
Price
6.684,16 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The Pipetting Head Height Adjuster is an accessory for the PLATEMASTER® that allows for easy altering of pipetting height, ensuring appropriate tip depth within shallow- or deep-well microplates.
The Lubrication Box of O-rings is an accessory for the PLATEMASTER®. We recommend lightly lubricating the external O-ring between regular maintenance intervals in situations of intensive use and to avoid ill-fitting tips.
Specifikationer
  • Beskrivelse:
    384-well plate adapter, positioning by hand