Accessories for PLATEMASTER®
Alu-heater block (96× 0,2 mm) adapter for non-skirted and semi-skirted plates
Katalog # 631-1372
Leverandør: GILSON
Gå til:
Product Details & Documents
The Pipetting Head Height Adjuster is an accessory for the PLATEMASTER® that allows for easy altering of pipetting height, ensuring appropriate tip depth within shallow- or deep-well microplates.
The Lubrication Box of O-rings is an accessory for the PLATEMASTER®. We recommend lightly lubricating the external O-ring between regular maintenance intervals in situations of intensive use and to avoid ill-fitting tips.
Documents
Specifikationer
- Beskrivelse:Alu-heater block (96× 0,2 mm) adapter for non-skirted and semi-skirted plates