Drawers, Transparent
2 l, transparent, 166×275×61 mm
MULC11102MU02000C
System of modular drawer cabinets made from PS. For well organised storage of many items, on shelves or at a workstation.
- Total visibility in the drawer
- Each drawer can be assembled horizontally and vertically
- Each drawer can divided with separators
Documents
VWR® For Your Space VWR® Lab bric-a-brac
Product Literature
- Kapacitet:2 l
- Farve:transparent
- Seq:02
- B×D×H:166×275×61 mm
Accessories
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse