Orders LinkContactUsLinkComponent
VWR®, LED-Fluorescence Cubes
VWR®, LED-Fluorescence Cubes
Far red LED fluorescence cube for TL534B FL, LED emission 740 nm,
Excitation filter 720 - 760 nm,
Dichroic mirror cut-off 770 nm,
Emission filter 780 LP nm
Catalog # 630-3252
Supplier:   VWR Collection
VWR®, LED-Fluorescence Cubes
Catalog # 630-3252
Supplier:  VWR Collection
VWR®, LED-Fluorescence Cubes
Far red LED fluorescence cube for TL534B FL, LED emission 740 nm,
Excitation filter 720 - 760 nm,
Dichroic mirror cut-off 770 nm,
Emission filter 780 LP nm
Price
kr29,320.00
Log in for availability
1 * 1 Items
Product Details & Documents
For use with microscope series TL534B FL.
Specifications
  • Beskrivelse:
    Far red LED fluorescence cube for TL534B FL
  • Optical features:
    LED emission 740 nm,
    Excitation filter 720 - 760 nm,
    Dichroic mirror cut-off 770 nm,
    Emission filter 780 LP nm