Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Microplate readers, Multiskan SkyHigh

Multiskan SkyHigh with touch screen, cuvette and µDrop Duo
  LABLA51119700DPC
Microplate readers, Multiskan SkyHigh
  LABLA51119700DPC
Microplate readers, Multiskan SkyHigh
Microplate readers, Multiskan SkyHigh

Multiskan SkyHigh with touch screen, cuvette and µDrop Duo

 
163.680,00 kr
   
1 * 1 Items
 

The Multiskan SkyHigh is a UV/Visible microplate spectrophotometer designed to be convenient and easy to use for virtually any photometric research application, especially DNA, RNA and protein analysis. It is ideal for multi-user environments where a variety of endpoint, kinetic and spectral assays are performed.

  • Fast, easy measurement - low volume nucleic acid or protein samples
  • Simple to use software - enables experiment set-up within minutes
  • Remote access - manage your data from anywhere via the Cloud, USB, or the Microsoft™ OneDrive™ platform

Multiskan Sky touch screen models can be connected to Thermo Fisher Connect cloud-based tools to enable secure digital data management, remote access of your instrument’s photometric data, and sharing between colleagues.

SkanIt™ software offers a comprehensive selection of built-in calculations, including blank subtraction, standard curves and concentration/dose-response curve fitting, classification, quality control, kinetic, and spectral analysis. Each user can install SkanIt software on his/her own computer to set up assays and analyse data. Thermo Fisher Connect cloud-based capabilities enable you to:
- Monitor the status of your instrument from your PC or Mac® computer
- Monitor the status of your instrument from your mobile device with the Instrument Connect app
- Upload measured data automatically or manually from your instrument to your cloud account
- Securely store, access, share, and manage your data remotely
- Receive automatic notification of new instrument software updates
- Upgrade instrument software at your convenience

 
  • Beskrivelse:
    Multiskan SkyHigh with touch screen, cuvette and µDrop Duo
  • Inkubationsområde:
    From ambient temperature to 45 ˚C
  • Valg af bølgelængde:
    Monochromator
  • B×D×H:
    295×445×265 mm
  • Måletid (96-brøndsplade):
    6 s
  • Linearitet:
    0 - 2,5 Abs, 2% (at 450 nm)
  • Scanningshastighed:
    10 s from 200 - 1000 nm in 1 nm steps
  • Båndbredde:
    <2,5 nm
  • Præcision:
    SD <0,003 Abs or CV <1,0% (at 450 nm)
  • Cat. No.:
    LABLA51119700DPC
  • Nøjagtighed:
    1,0% + 0,003 Abs (0 - 2,0 Abs), 2,0% (2,0 - 2,5 Abs)
  • Lyskilde:
    Xenon flash lamp
  • Område:
    Up to 4 Abs
  • Brugergrænseflade:
    Stand alone use: 7" touch screen display; PC control: Thermo Scientific™ SkanIt™ software
  • Måletid (384-brøndsplade):
    10 s
  • Vægt:
    11,3 kg
  • Rystning:
    Linear
  • Pladetyper:
    6- to 48-, 96-, and 384-well plates (max. height 19,5 mm with lid), Thermo Scientific™ µDrop™ Plates
  • Kuvettetype:
    W×D×H: 12,5×12,5×40 - 58 mm
    Beam centre height: 8,5 mm
    Beam window: ≥2
  • Bølgelængdeområde:
    200 - 1000 nm (1 nm steps)
  • Interface:
    1× USB B port for PC
    1× Ethernet port
    3× USB A ports for devices (USB memory device and Wi-Fi dongle)
Accessories

 

  •  
    Beskrivelse
     
     
  • 736-0754
    µDrop™ Plate, 16 measurement positions in a 2×8 matrix
       
    1 * 1 Items - 24.510,00 kr

 

  •  
    Beskrivelse
     
     
  • LABLN12391M2
    µDrop™ Duo plate, 2× 16 measurement positions in two 2×8 matrixes
       
    1 * 1 Items - 26.600,00 kr